Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001014431.1 | NP_001014431.1 | ||||
NM_001014432.1 | NP_001014432.1 | |||||
NM_005163.2 | NP_005154.2 | |||||
XM_005267401.1 | XP_005267458.1 | |||||
XM_017021075.1 | XP_016876564.1 | |||||
XM_017021076.1 | XP_016876565.1 | |||||
XM_017021077.1 | XP_016876566.1 | |||||
XM_017021078.1 | XP_016876567.1 | |||||
GenBank mRNA | AB451242.1 | - | ||||
AB451367.1 | - | |||||
AK094287.1 | - | |||||
AK122894.1 | - | |||||
AK131465.1 | - | |||||
AK299310.1 | - | |||||
AK314256.1 | - | |||||
BC000479.2 | - | 2964603 | ||||
BC001737.1 | - | |||||
BC063408.1 | - | |||||
BC084538.1 | - | 6645486 | ||||
BX647722.1 | - | |||||
BX648205.1 | - | |||||
CN414251.1 | - | |||||
ENST00000349310.0 | - | |||||
KF836747.1 | - | |||||
KF836748.1 | - | |||||
KF836749.1 | - | |||||
KF836750.1 | - | |||||
KU177898.1 | - | |||||
KU177899.1 | - | |||||
M63167.1 | - | |||||
X61037.1 | - |