Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000618.4 | NP_000609.1 | ||||
NM_001111283.2 | NP_001104753.1 | |||||
NM_001111284.1 | NP_001104754.1 | |||||
XM_017019259.1 | XP_016874748.1 | |||||
XM_017019260.1 | XP_016874749.1 | |||||
XM_017019261.1 | XP_016874750.1 | |||||
XM_017019262.1 | XP_016874751.1 | |||||
XM_017019263.1 | XP_016874752.1 | |||||
GenBank mRNA | AB209184.1 | - | ||||
AK312231.1 | - | |||||
AL603526.1 | - | |||||
AL710591.1 | - | |||||
BC148266.1 | - | 8860206 | ||||
BC152321.1 | - | 40129266 | ||||
CR541861.1 | - | |||||
ENST00000337514.0 | - | |||||
M11568.1 | - | |||||
M27544.1 | - | |||||
M29644.1 | - | |||||
M37484.1 | - | |||||
U40870.1 | - | |||||
X00173.1 | - | |||||
X56773.1 | - | |||||
X56774.1 | - | |||||
X57025.1 | - |