Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001118887.1 | NP_001112359.1 | ||||
NM_001118888.1 | NP_001112360.1 | |||||
NM_001147.2 | NP_001138.1 | |||||
XM_017013318.1 | XP_016868807.1 | |||||
GenBank mRNA | AB009865.1 | - | ||||
AF004327.1 | - | |||||
AF187858.1 | - | |||||
AF218015.1 | - | |||||
AJ289780.1 | - | |||||
AJ289781.1 | - | |||||
AK075219.1 | - | |||||
AK225698.1 | - | |||||
AK290070.1 | - | |||||
AK310171.1 | - | |||||
AK312541.1 | - | |||||
BC022490.1 | - | |||||
BC126200.1 | - | 8991916 | ||||
BC126202.1 | - | 8991918 | ||||
BC143902.1 | - | 9052420 | ||||
DA829327.1 | - | |||||
ENST00000325203.0 | - |