Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000882.3 | NP_000873.2 | ||||
GenBank mRNA | AF101062.1 | - | ||||
AF180562.1 | - | |||||
AW472810.1 | - | |||||
BC035571.1 | - | |||||
BC104982.1 | - | 8143985 | ||||
BC104984.1 | - | 8143987 | ||||
BG702253.1 | - | |||||
ENST00000305579.0 | - | |||||
M65271.1 | - | |||||
M65291.1 | - |