Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP2K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001316332.1 | NP_001303261.1 | ||||
NM_002756.4 | NP_002747.2 | |||||
NM_145109.2 | NP_659731.1 | |||||
XM_005256723.2 | XP_005256780.1 | |||||
XM_011523958.2 | XP_011522260.1 | |||||
XM_011523959.1 | XP_011522261.1 | |||||
XM_017024857.1 | XP_016880346.1 | |||||
XM_017024858.1 | XP_016880347.1 | |||||
XM_017024859.1 | XP_016880348.1 | |||||
GenBank mRNA | AB451226.1 | - | ||||
AB451349.1 | - | |||||
AI758302.1 | - | |||||
AK093838.1 | - | |||||
AK223490.1 | - | |||||
AK315505.1 | - | |||||
BC032478.1 | - | 5215093 | ||||
BM836867.1 | - | |||||
CR536514.1 | - | |||||
CR541793.1 | - | |||||
D87116.1 | - | |||||
DA131175.1 | - | |||||
DA987047.1 | - | |||||
ENST00000361818.0 | - | |||||
GQ225578.1 | - | |||||
L36719.1 | - | |||||
U66839.1 | - |