Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001258206.1 | NP_001245135.1 | ||||
NM_001258207.1 | NP_001245136.1 | |||||
NM_005428.3 | NP_005419.2 | |||||
XM_005259642.1 | XP_005259699.1 | |||||
GenBank mRNA | AK225710.1 | - | ||||
AK301128.1 | - | |||||
AK313307.1 | - | |||||
AW105491.1 | - | |||||
BC013361.2 | - | 3842829 | ||||
DC408645.1 | - | |||||
ENST00000304076.0 | - | |||||
X16316.1 | - | |||||
X83931.1 | - |