Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKBKE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001193321.1 | NP_001180250.1 | ||||
NM_001193322.1 | NP_001180251.1 | |||||
NM_014002.3 | NP_054721.1 | |||||
XM_005273356.2 | XP_005273413.1 | |||||
XM_017002868.1 | XP_016858357.1 | |||||
GenBank mRNA | AB016590.1 | - | ||||
AF241789.1 | - | |||||
AK093798.1 | - | |||||
AK310737.1 | - | |||||
AL706097.1 | - | |||||
AY660028.1 | - | |||||
BC105923.1 | - | 40001562 | ||||
BC105924.1 | - | 40001567 | ||||
BC107591.1 | - | |||||
BC107812.1 | - | 40001564 | ||||
CB243308.1 | - | |||||
D63485.1 | - | |||||
DA990116.1 | - | |||||
DA999897.1 | - | |||||
DB152295.1 | - | |||||
ENST00000367120.0 | - |