Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_019111.4 | NP_061984.2 | ||||
GenBank mRNA | AI492718.1 | - | ||||
AK297032.1 | - | |||||
AK301134.1 | - | |||||
AK301190.1 | - | |||||
AK301354.1 | - | |||||
AK304429.1 | - | |||||
AK310161.1 | - | |||||
AK313123.1 | - | |||||
BC032350.1 | - | 5214156 | ||||
BC071659.1 | - | 5162925 | ||||
BG548634.1 | - | |||||
BM849755.1 | - | |||||
CR457013.1 | - | |||||
ENST00000395388.0 | - | |||||
J00193.1 | - | |||||
J00194.1 | - | |||||
J00195.1 | - | |||||
J00197.1 | - | |||||
K01171.1 | - | |||||
M35979.1 | - | |||||
M60333.1 | - | |||||
M60334.1 | - | |||||
V00523.1 | - | |||||
V00528.1 | - |