Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002401.3 | NP_002392.2 | ||||
NM_203351.1 | NP_976226.1 | |||||
XM_005257376.3 | XP_005257433.1 | |||||
XM_005257377.3 | XP_005257434.1 | |||||
XM_005257378.2 | XP_005257435.1 | |||||
GenBank mRNA | AF172823.1 | - | ||||
AK022333.1 | - | |||||
AK225784.1 | - | |||||
AK308152.1 | - | |||||
AK315305.1 | - | |||||
AL834303.1 | - | |||||
AX775739.1 | - | |||||
AX775741.1 | - | |||||
BC008336.1 | - | |||||
BC010464.1 | - | |||||
BC018021.1 | - | |||||
BC090859.1 | - | 6183752 | ||||
BC093672.1 | - | 7939517 | ||||
BC093674.1 | - | 7939519 | ||||
BC143224.1 | - | |||||
BC143235.1 | - | |||||
BM479726.1 | - | |||||
BM728104.1 | - | |||||
BM800091.1 | - | |||||
BQ643432.1 | - | |||||
BU633068.1 | - | |||||
DQ454151.1 | - | |||||
ENST00000361733.0 | - | |||||
U78876.1 | - |