Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002264.3 | NP_002255.3 | ||||
NR_026698.1 | ||||||
XM_005247437.3 | XP_005247494.1 | |||||
XM_005247439.3 | XP_005247496.1 | |||||
XM_011512795.2 | XP_011511097.1 | |||||
XM_011512797.2 | XP_011511099.1 | |||||
XM_017006363.1 | XP_016861852.1 | |||||
XM_017006364.1 | XP_016861853.1 | |||||
XM_017006365.1 | XP_016861854.1 | |||||
GenBank mRNA | AA689229.1 | - | ||||
AF035311.1 | - | |||||
AI216375.1 | - | |||||
AK125626.1 | - | |||||
AK128280.1 | - | |||||
AK299030.1 | - | |||||
AK312713.1 | - | |||||
AL700464.1 | - | |||||
AL710311.1 | - | |||||
BC002374.2 | - | 2822859 | ||||
BC003009.1 | - | 2822859 | ||||
BC029937.1 | - | |||||
BC090864.1 | - | 6173605 | ||||
BG287593.1 | - | |||||
BT006959.1 | - | |||||
CA433573.1 | - | |||||
CR456743.1 | - | |||||
DA308391.1 | - | |||||
DN991078.1 | - | |||||
ENST00000344337.0 | - | |||||
S75295.1 | - | |||||
U20620.1 | - |