Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_032965.5 | NP_116741.2 | ||||
GenBank mRNA | AF031587.1 | - | ||||
AI244164.1 | - | |||||
BX098368.1 | - | |||||
BX497188.1 | - | |||||
U58914.1 | - |