Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_144719.3 | NP_653320.3 | ||||
XM_005264898.3 | XP_005264955.1 | |||||
XM_005264899.4 | XP_005264956.1 | |||||
XM_011533418.2 | XP_011531720.1 | |||||
XM_011533419.2 | XP_011531721.1 | |||||
XM_011533420.2 | XP_011531722.1 | |||||
XM_011533421.2 | XP_011531723.1 | |||||
XM_017005781.1 | XP_016861270.1 | |||||
GenBank mRNA | AK058196.1 | - | ||||
AK302859.1 | - | |||||
BC036050.1 | - | 5270468 | ||||
BM664398.1 | - | |||||
ENST00000310232.0 | - |