Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002176.3 | NP_002167.1 | ||||
GenBank mRNA | AB451323.1 | - | ||||
AB451452.1 | - | |||||
AB451491.1 | - | |||||
BC069314.1 | - | 7262165 | ||||
BC096150.3 | - | 40002940 | ||||
BC096151.2 | - | 40002941 | ||||
BC096152.2 | - | 40002943 | ||||
BC096153.2 | - | 40002945 | ||||
ENST00000380232.0 | - | |||||
M17373.2 | - | |||||
M25460.1 | - | |||||
M28622.1 | - | |||||
V00546.1 | - | |||||
V00547.1 | - |