Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000948.5 | NP_000939.1 | ||||
NM_001163558.2 | NP_001157030.1 | |||||
XM_011514753.2 | XP_011513055.1 | |||||
XM_011514754.2 | XP_011513056.1 | |||||
GenBank mRNA | BC015850.1 | - | 4657724 | |||
BC088370.1 | - | 30393428 | ||||
CD106243.1 | - | |||||
D00411.1 | - | |||||
DQ839492.1 | - | |||||
ENST00000306482.0 | - | |||||
GQ305133.1 | - | |||||
M29386.1 | - | |||||
M58594.1 | - | |||||
U75583.1 | - | |||||
V00566.1 | - | |||||
X54393.1 | - | |||||
X54394.1 | - |