Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001278599.1 | NP_001265528.1 | ||||
NM_001699.5 | NP_001690.2 | |||||
NM_021913.4 | NP_068713.2 | |||||
GenBank mRNA | AA021223.1 | - | ||||
AI300772.1 | - | |||||
AI309979.1 | - | |||||
AK309139.1 | - | |||||
AK310881.1 | - | |||||
AL556721.3 | - | |||||
BC032229.1 | - | 5205825 | ||||
CA437238.1 | - | |||||
DA478638.1 | - | |||||
ENST00000359092.0 | - | |||||
HY045208.1 | - | |||||
M76125.1 | - | |||||
S65125.1 | - | |||||
X57019.1 | - | |||||
X66029.1 | - |