Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002506.2 | NP_002497.2 | ||||
XM_006710663.3 | XP_006710726.1 | |||||
XM_011541518.2 | XP_011539820.1 | |||||
GenBank mRNA | AF411526.1 | - | ||||
BC032517.2 | - | |||||
BC126148.1 | - | 8991864 | ||||
BC126150.1 | - | 8991866 | ||||
BE045524.1 | - | |||||
BT019733.1 | - | |||||
CR541855.1 | - | |||||
ENST00000369512.0 | - | |||||
X52599.1 | - |