Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001289160.1 | NP_001276089.1 | ||||
NM_001289161.1 | NP_001276090.1 | |||||
NM_005931.4 | NP_005922.2 | |||||
GenBank mRNA | AF021221.1 | - | ||||
AF021222.1 | - | |||||
AF021223.1 | - | |||||
AF021224.1 | - | |||||
AF021225.1 | - | |||||
AF021226.1 | - | |||||
AK291927.1 | - | |||||
AK300791.1 | - | |||||
AK303655.1 | - | |||||
AK314228.1 | - | |||||
AY885251.1 | - | |||||
BC044218.1 | - | 6025253 | ||||
BQ216247.1 | - | |||||
BU684700.1 | - | |||||
CN389975.1 | - | |||||
DB121254.1 | - | |||||
DC364717.1 | - | |||||
DC427936.1 | - | |||||
ENST00000252229.0 | - | |||||
GU235997.1 | - | |||||
U95729.1 | - | |||||
U95730.1 | - | |||||
U95731.1 | - | |||||
U95732.1 | - | |||||
U95733.1 | - | |||||
U95734.1 | - | |||||
X91625.1 | - |