Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKBKG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001099856.4 | NP_001093326.2 | ||||
NM_001099857.2 | NP_001093327.1 | |||||
NM_001145255.2 | NP_001138727.1 | |||||
NM_001321396.1 | NP_001308325.1 | |||||
NM_001321397.1 | NP_001308326.1 | |||||
NM_003639.4 | NP_003630.1 | |||||
GenBank mRNA | AF062089.1 | - | ||||
AF074382.1 | - | |||||
AF091453.1 | - | |||||
AF261086.1 | - | |||||
AK000593.1 | - | |||||
AK308366.1 | - | |||||
AY114157.1 | - | |||||
AY429546.1 | - | |||||
AY429547.1 | - | |||||
AY429548.1 | - | |||||
BC000299.2 | - | 2820134 | ||||
BC012114.1 | - | 4651870 | ||||
BC046922.1 | - | 5476394 | ||||
BC050612.2 | - | 6062527 | ||||
BM473416.1 | - | |||||
BT019621.1 | - | |||||
CA916723.1 | - | |||||
DC410130.1 | - |