Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_005408.2 | NP_005399.1 | ||||
GenBank mRNA | AJ001634.1 | - | ||||
BC008621.1 | - | 4184250 | ||||
BT007385.1 | - | |||||
CR450337.1 | - | |||||
ENST00000225844.0 | - | |||||
U46767.1 | - | |||||
U59808.1 | - | |||||
X98306.1 | - | |||||
Z77650.1 | - | |||||
Z77651.1 | - |