Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NR_033351.1 | |||||
GenBank mRNA | AK057241.1 | - | ||||
AK302625.1 | - | |||||
AL833871.1 | - | |||||
BC144672.1 | - | |||||
BC150302.1 | - | 8860399 | ||||
DB071191.1 | - | |||||
DQ309038.1 | - | |||||
ENST00000532622.0 | - |