Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000389.4 | NP_000380.1 | ||||
NM_001220777.1 | NP_001207706.1 | |||||
NM_001220778.1 | NP_001207707.1 | |||||
NM_001291549.1 | NP_001278478.1 | |||||
NM_078467.2 | NP_510867.1 | |||||
GenBank mRNA | AA534613.1 | - | ||||
AB209881.1 | - | |||||
AB451290.1 | - | |||||
AB451422.1 | - | |||||
AI973138.1 | - | |||||
AK298901.1 | - | |||||
AK309512.1 | - | |||||
BC000275.1 | - | 3355833 | ||||
BC000312.2 | - | 2821049 | ||||
BC001935.1 | - | 2823668 | ||||
BC013967.2 | - | 2822909 | ||||
BQ941282.1 | - | |||||
BT006719.1 | - | |||||
BU154329.1 | - | |||||
BU193165.1 | - | |||||
CR536533.1 | - | |||||
DA552678.1 | - | |||||
DA582789.1 | - | |||||
DA699672.1 | - | |||||
DA702185.1 | - | |||||
DA838987.1 | - | |||||
ENST00000244741.0 | - | |||||
L25610.1 | - | |||||
L26165.1 | - | |||||
L47232.1 | - | |||||
L47233.1 | - | |||||
S67388.1 | - | |||||
U03106.1 | - | |||||
U09579.1 | - |