Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002745.4 | NP_002736.3 | ||||
NM_138957.3 | NP_620407.1 | |||||
GenBank mRNA | AI004470.1 | - | ||||
AK055080.1 | - | |||||
AK057143.1 | - | |||||
AK090869.1 | - | |||||
AK295188.1 | - | |||||
AK307615.1 | - | |||||
BC017832.1 | - | 4685557 | ||||
BC099905.1 | - | 5264558 | ||||
BQ774391.1 | - | |||||
DQ399292.1 | - | |||||
EF565108.1 | - | |||||
ENST00000215832.0 | - | |||||
Z11694.1 | - | |||||
Z11695.1 | - |