Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001251825.1 | NP_001238754.1 | ||||
NM_003109.1 | NP_003100.1 | |||||
NM_138473.2 | NP_612482.2 | |||||
XM_011538696.2 | XP_011536998.1 | |||||
GenBank mRNA | AB209678.1 | - | ||||
AF252284.1 | - | |||||
AF255682.1 | - | |||||
AI282745.1 | - | |||||
AI561005.1 | - | |||||
AJ272134.1 | - | |||||
AK311995.1 | - | |||||
AL442093.1 | - | |||||
AW028976.1 | - | |||||
BC012008.1 | - | |||||
BC043224.1 | - | |||||
BC062539.1 | - | 5928633 | ||||
BC078158.1 | - | |||||
BF434304.1 | - | |||||
BF436290.1 | - | |||||
BF671559.1 | - | |||||
BM463205.1 | - | |||||
BQ072120.1 | - | |||||
BQ431320.1 | - | |||||
BQ774060.1 | - | |||||
CD104746.1 | - | |||||
FJ949572.1 | - | |||||
FN908228.1 | - | |||||
J03133.1 | - |