Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_005429.4 | NP_005420.1 | ||||
GenBank mRNA | AK313879.1 | - | ||||
AW294939.1 | - | |||||
BC035212.1 | - | 4753457 | ||||
BC063685.1 | - | 4513934 | ||||
BF528469.1 | - | |||||
CR541897.1 | - | |||||
DA044965.1 | - | |||||
U43142.1 | - | |||||
U58111.1 | - | |||||
X94216.1 | - |