Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001201359.1 | NP_001188288.1 | ||||
NM_005624.3 | NP_005615.2 | |||||
XM_011528177.2 | XP_011526479.1 | |||||
XM_017027119.1 | XP_016882608.1 | |||||
XM_017027120.1 | XP_016882609.1 | |||||
XM_017027121.1 | XP_016882610.1 | |||||
XM_017027122.1 | XP_016882611.1 | |||||
GenBank mRNA | AB046579.1 | - | ||||
AI313430.1 | - | |||||
AI961745.1 | - | |||||
AK292662.1 | - | |||||
BC125136.1 | - | 40119547 | ||||
BC130561.1 | - | 40146592 | ||||
BC144463.1 | - | 9052989 | ||||
ENST00000390669.0 | - | |||||
U86358.1 | - |