Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000247.2 | NP_000238.1 | ||||
NM_001177519.2 | NP_001170990.1 | |||||
NM_001289152.1 | NP_001276081.1 | |||||
NM_001289153.1 | NP_001276082.1 | |||||
NM_001289154.1 | NP_001276083.1 | |||||
GenBank mRNA | AB506764.1 | - | ||||
AK094237.1 | - | |||||
AK225393.1 | - | |||||
AK299135.1 | - | |||||
AK310028.1 | - | |||||
AK311667.1 | - | |||||
AY204547.1 | - | |||||
AY750850.1 | - | |||||
BC016929.1 | - | 4446575 | ||||
BM552351.1 | - | |||||
BM991681.1 | - | |||||
BX342160.2 | - | |||||
CN483948.1 | - | |||||
DT219772.1 | - | |||||
ENST00000364810.0 | - | |||||
L14848.1 | - |