Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001079.3 | NP_001070.2 | ||||
NM_207519.1 | NP_997402.1 | |||||
XM_017004867.1 | XP_016860356.1 | |||||
XM_017004868.1 | XP_016860357.1 | |||||
XM_017004869.1 | XP_016860358.1 | |||||
XM_017004870.1 | XP_016860359.1 | |||||
GenBank mRNA | AB083211.1 | - | ||||
AK303341.1 | - | |||||
AK310628.1 | - | |||||
AK310637.1 | - | |||||
AL832287.1 | - | |||||
BC039039.1 | - | 5168073 | ||||
BC053878.1 | - | 5757161 | ||||
ENST00000264972.0 | - | |||||
L05148.1 | - |