Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002507.3 | NP_002498.1 | ||||
GenBank mRNA | AK303278.1 | - | ||||
AK313654.1 | - | |||||
BC050309.1 | - | 5263715 | ||||
BC052797.1 | - | |||||
BQ639556.1 | - | |||||
CK299927.1 | - | |||||
DA115173.1 | - | |||||
ENST00000172229.0 | - | |||||
M14764.1 | - |