Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001256560.1 | NP_001243489.1 | ||||
NM_004944.3 | NP_004935.1 | |||||
GenBank mRNA | AB209613.1 | - | ||||
AF047354.1 | - | |||||
AI591302.1 | - | |||||
AI660448.1 | - | |||||
AK131323.1 | - | |||||
AK301263.1 | - | |||||
AK313303.1 | - | |||||
BC015831.1 | - | 4723725 | ||||
BI521582.1 | - | |||||
BI834035.1 | - | |||||
DB164811.1 | - | |||||
ENST00000394549.0 | - | |||||
U56814.1 | - |