Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000515.4 | NP_000506.2 | ||||
NM_022559.3 | NP_072053.1 | |||||
NM_022560.3 | NP_072054.1 | |||||
GenBank mRNA | A00469.1 | - | ||||
AF110644.1 | - | |||||
AF185611.1 | - | |||||
AV749353.1 | - | |||||
AV753220.1 | - | |||||
AY613431.1 | - | |||||
AY613432.1 | - | |||||
BC062475.1 | - | 30393581 | ||||
BC075012.2 | - | 30915404 | ||||
BC075013.2 | - | 30915617 | ||||
BC090045.1 | - | 30394115 | ||||
CD108592.1 | - | |||||
ENST00000323322.0 | - | |||||
M14398.1 | - | |||||
V00519.1 | - |