Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFHAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_004225.2 | NP_004216.2 | ||||
XM_011543852.2 | XP_011542154.1 | |||||
GenBank mRNA | AB016816.1 | - | ||||
AK027007.1 | - | |||||
AK096504.1 | - | |||||
BC014226.2 | - | |||||
ENST00000276282.0 | - |