Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD36BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NR_015424.1 | |||||
GenBank mRNA | AK097649.1 | - | ||||
AL137717.1 | - | |||||
BC031698.1 | - | |||||
BX537549.1 | - | |||||
BX648045.1 | - |