Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL6R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000565.3 | NP_000556.1 | ||||
NM_001206866.1 | NP_001193795.1 | |||||
NM_181359.2 | NP_852004.1 | |||||
XM_005245139.1 | XP_005245196.1 | |||||
XM_005245140.2 | XP_005245197.1 | |||||
XM_006711298.1 | XP_006711361.1 | |||||
XM_006711299.3 | XP_006711362.1 | |||||
XM_017001199.1 | XP_016856688.1 | |||||
XM_017001200.1 | XP_016856689.1 | |||||
XM_017001201.1 | XP_016856690.1 | |||||
GenBank mRNA | AA035377.1 | - | ||||
AK223582.1 | - | |||||
AK293013.1 | - | |||||
AK312730.1 | - | |||||
BC035327.1 | - | |||||
BC089410.1 | - | 3078506 | ||||
BC132684.1 | - | 40146706 | ||||
BC132686.1 | - | 40146708 | ||||
BM911928.1 | - | |||||
S72848.1 | - | |||||
X12830.1 | - | |||||
X58298.1 | - |