Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002020.4 | NP_002011.2 | ||||
NM_182925.4 | NP_891555.2 | |||||
XM_011534478.2 | XP_011532780.1 | |||||
XM_011534484.2 | XP_011532786.1 | |||||
XM_017009263.1 | XP_016864752.1 | |||||
XM_017009264.1 | XP_016864753.1 | |||||
XM_017009265.1 | XP_016864754.1 | |||||
XM_017009266.1 | XP_016864755.1 | |||||
XM_017009267.1 | XP_016864756.1 | |||||
XM_017009268.1 | XP_016864757.1 | |||||
GenBank mRNA | AB209637.1 | - | ||||
AK291679.1 | - | |||||
AK309910.1 | - | |||||
AY233382.1 | - | |||||
AY233383.1 | - | |||||
BC027302.1 | - | |||||
CA438330.1 | - | |||||
DB455035.2 | - | |||||
ENST00000393347.0 | - | |||||
EU826564.1 | - | |||||
EU826565.1 | - | |||||
EU826566.1 | - | |||||
S66407.1 | - | |||||
U43143.1 | - | |||||
X68203.1 | - | |||||
X69878.1 | - |