Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_005228.3 | NP_005219.2 | ||||
NM_201282.1 | NP_958439.1 | |||||
NM_201283.1 | NP_958440.1 | |||||
NM_201284.1 | NP_958441.1 | |||||
GenBank mRNA | AB209442.1 | - | ||||
AF125253.1 | - | |||||
AF277897.1 | - | |||||
AI217671.1 | - | |||||
AK127817.1 | - | |||||
AK225422.1 | - | |||||
AK290352.1 | - | |||||
AK294750.1 | - | |||||
AL598260.1 | - | |||||
AU137334.1 | - | |||||
AW163038.1 | - | |||||
AW295229.1 | - | |||||
AY550966.1 | - | |||||
AY698023.1 | - | |||||
AY698024.1 | - | |||||
BC070081.1 | - | |||||
BC094761.1 | - | 30528231 | ||||
BC118665.1 | - | |||||
BC128419.1 | - | |||||
CB160831.1 | - | |||||
DQ088980.1 | - | |||||
EF210210.1 | - | |||||
EF210211.1 | - | |||||
ENST00000275493.0 | - | |||||
GU228573.1 | - | |||||
GU228584.1 | - | |||||
GU255993.1 | - | |||||
HM437234.1 | - | |||||
HM437235.1 | - | |||||
HQ730875.1 | - | |||||
HQ912715.1 | - | |||||
HQ999997.1 | - | |||||
K03193.1 | - | |||||
U48722.1 | - | |||||
U51732.1 | - | |||||
U95089.1 | - | |||||
X00588.1 | - | |||||
X00663.1 | - |