Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000314.6 | NP_000305.3 | ||||
NM_001304717.2 | NP_001291646.2 | |||||
NM_001304718.1 | NP_001291647.1 | |||||
GenBank mRNA | AA017584.1 | - | ||||
AA836562.1 | - | |||||
AI825848.1 | - | |||||
AK021619.1 | - | |||||
AK024986.1 | - | |||||
AK124525.1 | - | |||||
AK130076.1 | - | |||||
AK313581.1 | - | |||||
BC005821.2 | - | 3937787 | ||||
BG772190.1 | - | |||||
CA944942.1 | - | |||||
CR450306.1 | - | |||||
ENST00000371953.0 | - | |||||
JF268690.1 | - | |||||
KJ534922.1 | - | |||||
U92436.1 | - | |||||
U93051.1 | - | |||||
U96180.1 | - |