Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002944.2 | NP_002935.2 | ||||
XM_006715548.3 | XP_006715611.1 | |||||
XM_011536049.2 | XP_011534351.1 | |||||
XM_011536050.2 | XP_011534352.1 | |||||
XM_011536051.2 | XP_011534353.1 | |||||
XM_011536052.2 | XP_011534354.1 | |||||
XM_011536053.2 | XP_011534355.1 | |||||
XM_011536054.2 | XP_011534356.1 | |||||
XM_011536055.2 | XP_011534357.1 | |||||
XM_011536056.2 | XP_011534358.1 | |||||
XM_011536057.2 | XP_011534359.1 | |||||
XM_011536058.2 | XP_011534360.1 | |||||
XM_017011172.1 | XP_016866661.1 | |||||
XM_017011173.1 | XP_016866662.1 | |||||
GenBank mRNA | BC144556.1 | - | ||||
ENST00000368508.0 | - | |||||
M13880.1 | - | |||||
M34353.1 | - |