Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001200.3 | NP_001191.1 | ||||
XM_011529323.2 | XP_011527625.1 | |||||
GenBank mRNA | AA489383.1 | - | ||||
BC069214.1 | - | |||||
BM670141.1 | - | |||||
ENST00000378827.0 | - | |||||
KC294426.1 | - | |||||
KF250425.1 | - | |||||
M22489.1 | - |