Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001007792.1 | NP_001007793.1 | ||||
NM_001012331.1 | NP_001012331.1 | |||||
NM_002529.3 | NP_002520.2 | |||||
GenBank mRNA | AK126428.1 | - | ||||
AK290759.1 | - | |||||
AK312704.1 | - | |||||
BC062580.1 | - | |||||
BC136554.1 | - | 9020544 | ||||
BC144239.1 | - | 9052759 | ||||
BM685020.1 | - | |||||
CR936794.1 | - | |||||
ENST00000524377.0 | - | |||||
KJ534902.1 | - | |||||
KJ534903.1 | - | |||||
M23102.1 | - |