Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001229.4 | NP_001220.2 | ||||
NM_001278054.1 | NP_001264983.1 | |||||
NM_032996.3 | NP_127463.2 | |||||
NR_102732.1 | ||||||
NR_102733.1 | ||||||
XM_005246014.2 | XP_005246071.1 | |||||
XM_011542273.2 | XP_011540575.1 | |||||
GenBank mRNA | AA534393.1 | - | ||||
AB015653.1 | - | |||||
AB020979.1 | - | |||||
AB209147.1 | - | |||||
AF093130.1 | - | |||||
AF110376.1 | - | |||||
AJ781267.1 | - | |||||
AK292080.1 | - | |||||
AK292111.1 | - | |||||
AK303743.1 | - | |||||
AY732490.2 | - | |||||
BC002452.2 | - | 3343958 | ||||
BC006463.1 | - | 3542075 | ||||
BT006911.1 | - | |||||
DA078665.1 | - | |||||
ENST00000333868.0 | - | |||||
HY027003.1 | - | |||||
U56390.1 | - | |||||
U60521.1 | - |