Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001198557.1 | NP_001185486.1 | ||||
NM_005573.3 | NP_005564.1 | |||||
NR_134488.1 | ||||||
GenBank mRNA | AK303084.1 | - | ||||
AK312603.1 | - | |||||
AU134245.1 | - | |||||
AW512433.1 | - | |||||
BC012295.1 | - | 3445475 | ||||
BC052951.1 | - | |||||
BC078178.1 | - | |||||
BC103723.1 | - | 6146212 | ||||
ENST00000261366.0 | - | |||||
M34458.1 | - |