Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001099691.2 | NP_001093161.1 | ||||
NM_001308158.1 | NP_001295087.1 | |||||
NM_001308159.1 | NP_001295088.1 | |||||
NM_003236.3 | NP_003227.1 | |||||
GenBank mRNA | AF149096.1 | - | ||||
AF149097.1 | - | |||||
AF149098.1 | - | |||||
AK290151.1 | - | |||||
AK309066.1 | - | |||||
AK309220.1 | - | |||||
AK312899.1 | - | |||||
AY548763.1 | - | |||||
BC005308.1 | - | 3828239 | ||||
BI760767.1 | - | |||||
BM453873.1 | - | |||||
BT006833.1 | - | |||||
CD369689.1 | - | |||||
CN274505.1 | - | |||||
DA216480.1 | - | |||||
DA229819.1 | - | |||||
DA373586.1 | - | |||||
DA462271.1 | - | |||||
ENST00000295400.0 | - | |||||
K03222.1 | - | |||||
M31172.1 | - | |||||
X70340.1 | - |