Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000016.5 | NP_000007.1 | ||||
NM_001127328.2 | NP_001120800.1 | |||||
NM_001286042.1 | NP_001272971.1 | |||||
NM_001286043.1 | NP_001272972.1 | |||||
NM_001286044.1 | NP_001272973.1 | |||||
GenBank mRNA | AF251043.1 | - | ||||
AK296045.1 | - | |||||
AK301005.1 | - | |||||
AK301039.1 | - | |||||
AK301717.1 | - | |||||
AK309843.1 | - | |||||
AK311022.1 | - | |||||
AK311185.1 | - | |||||
AK312629.1 | - | |||||
AK315946.1 | - | |||||
BC005377.1 | - | 3934299 | ||||
BI789086.1 | - | |||||
BX647734.1 | - | |||||
DB055575.1 | - | |||||
DB453314.1 | - | |||||
DB537134.1 | - | |||||
ENST00000370841.0 | - | |||||
KU177876.1 | - | |||||
KU177877.1 | - | |||||
KU177878.1 | - | |||||
M16827.1 | - |