Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001135044.1 | NP_001128516.1 | ||||
NM_001308244.1 | NP_001295173.1 | |||||
NM_002752.4 | NP_002743.3 | |||||
NM_139068.2 | NP_620707.1 | |||||
NM_139069.2 | NP_620708.1 | |||||
NM_139070.2 | NP_620709.1 | |||||
XM_005265940.4 | XP_005265997.1 | |||||
XM_006714891.2 | XP_006714954.1 | |||||
XM_017009638.1 | XP_016865127.1 | |||||
XM_017009639.1 | XP_016865128.1 | |||||
XM_017009640.1 | XP_016865129.1 | |||||
XM_017009641.1 | XP_016865130.1 | |||||
XM_017009642.1 | XP_016865131.1 | |||||
XM_017009643.1 | XP_016865132.1 | |||||
GenBank mRNA | AB451302.1 | - | ||||
AB451355.1 | - | |||||
AB451433.1 | - | |||||
AK289638.1 | - | |||||
BC032539.1 | - | 5528624 | ||||
BF056535.1 | - | |||||
BQ025589.1 | - | |||||
CR536580.1 | - | |||||
CX758487.1 | - | |||||
DB451562.1 | - | |||||
ENST00000393360.0 | - | |||||
EU927388.1 | - | |||||
HM036706.1 | - | |||||
HM036707.1 | - | |||||
HM036708.1 | - | |||||
HM036709.1 | - | |||||
L31951.1 | - | |||||
U09759.1 | - | |||||
U34821.1 | - | |||||
U35002.1 | - | |||||
U35003.1 | - |