Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_003844.3 | NP_003835.3 | ||||
GenBank mRNA | AK291299.1 | - | ||||
BC012866.1 | - | 3857315 | ||||
BT006906.1 | - | |||||
CN423254.1 | - | |||||
DB101031.1 | - | |||||
ENST00000221132.0 | - | |||||
FJ795027.1 | - | |||||
JF729355.1 | - | |||||
U90875.1 | - |