Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001206389.1 | NP_001193318.1 | ||||
NM_006119.4 | NP_006110.1 | |||||
NM_033163.3 | NP_149353.1 | |||||
NM_033164.3 | NP_149354.1 | |||||
NM_033165.3 | NP_149355.1 | |||||
GenBank mRNA | AB014615.1 | - | ||||
BC069106.1 | - | 7216922 | ||||
BC128235.1 | - | 40114671 | ||||
BC128236.1 | - | 40114672 | ||||
CN369230.1 | - | |||||
ENST00000320185.0 | - | |||||
U36223.1 | - | |||||
U46211.1 | - | |||||
U46212.1 | - | |||||
U46213.1 | - | |||||
U56978.1 | - |