Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_004465.1 | NP_004456.1 | ||||
XM_005248264.3 | XP_005248321.1 | |||||
GenBank mRNA | AB002097.1 | - | ||||
AF411527.1 | - | |||||
AF508782.1 | - | |||||
AK023508.1 | - | |||||
AK315314.1 | - | |||||
BC069561.1 | - | 7262177 | ||||
BC105021.1 | - | 8144024 | ||||
BC105023.1 | - | 8144026 | ||||
BC143772.1 | - | |||||
CR541665.1 | - | |||||
CR541688.1 | - | |||||
ENST00000264664.0 | - | |||||
GQ351295.1 | - | |||||
U67918.1 | - |