Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_006179.4 | NP_006170.1 | ||||
XM_005258962.3 | XP_005259019.1 | |||||
XM_006723232.3 | XP_006723295.1 | |||||
XM_011527008.2 | XP_011525310.1 | |||||
XM_011527009.2 | XP_011525311.1 | |||||
XM_011527010.2 | XP_011525312.1 | |||||
GenBank mRNA | AK297906.1 | - | ||||
BC012421.2 | - | 3865300 | ||||
BT019368.1 | - | |||||
BT019369.1 | - | |||||
CR541900.1 | - | |||||
ENST00000301411.0 | - |