Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001009552.1 | NP_001009552.1 | ||||
GenBank mRNA | AF444006.1 | - | ||||
AK091875.1 | - | |||||
BC012022.1 | - | 4454972 | ||||
BG716837.1 | - | |||||
BT019526.1 | - | |||||
CA419001.1 | - | |||||
CF594136.1 | - | |||||
CR541747.1 | - | |||||
ENST00000221138.0 | - | |||||
J03805.1 | - | |||||
X12656.1 | - |