Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_005623.2 | NP_005614.2 | ||||
GenBank mRNA | BC126242.1 | - | 8991958 | |||
ENST00000394620.0 | - | |||||
Y10802.1 | - | |||||
Y16645.1 | - |